Learn More
Medchemexpress LLC 3-(6-methyl-2-pyridinyl)-N-phenyl-4-quinolin-4-ylpyrazole-1-carbothioamide | 909910-43-6 | MFCD08705403 | >98.0% | 421.52 g/mol | C25H19N5S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10432G25MG
Encompass_Preferred
A 83-01 (GMP) is the GMP-grade form of A-83-01, a potent small-molecule inhibitor of activin receptor-like kinases ALK4, ALK5 (TGF-β type I), and ALK7. Supplied for research and cell therapy manufacturing as an auxiliary reagent produced under GMP guidelines, the compound (CAS 909910-43-6) is used to block TGF-β signaling in cell-based assays and differentiation protocols.
  • Potent inhibition of ALK4, ALK5, and ALK7 for TGF-β pathway modulation.
  • GMP-grade material suitable for cell therapy manufacturing workflows.
  • High purity, ≥98% (HPLC), for consistent experimental results.
  • Solid small molecule with formula C25H19N5S and molecular weight 421.52 g/mol.
  • Suitable as an auxiliary reagent in cell culture and signaling studies.
Catalog No. 50-002-42603