Learn More
Medchemexpress LLC 3-(6-methyl-2-pyridinyl)-N-phenyl-4-quinolin-4-ylpyrazole-1-carbothioamide | 909910-43-6 | MFCD08705403 | 98.7% | 421.52 g/mol | C25H19N5S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY104321MG
Encompass_Preferred
A 83-01 is a small-molecule research standard that potently inhibits transforming growth factor-β (TGF-β) type I receptor kinases, including ALK5, ALK4, and ALK7. It is supplied as a solid analytical standard for biochemical and cell-based studies, with confirmed molecular formula C25H19N5S and high reported purity suitable for research applications.
  • Potent inhibition of TGF-β type I receptor signaling.
  • Useful in biochemical and cellular assays for pathway modulation.
  • High reported purity for analytical reliability.
  • Solid, stable form suitable for storage and handling.
  • Documented physicochemical data including formula and molecular weight.
Catalog No. 50-002-42823