Learn More
Medchemexpress LLC 3-(6-methyl-2-pyridinyl)-N-phenyl-4-quinolin-4-ylpyrazole-1-carbothioamide | 909910-43-6 | MFCD08705403 | 99.0% | 421.52 g·mol⁻¹ | C25H19N5S | 5 MG

Supplier: Medchemexpress LLC HY10432R5MG
A 83-01 is a small-molecule inhibitor of TGF-β type I receptor kinases (ALK4, ALK5, and ALK7) used in biochemical and cellular research to block activin/TGF-β signaling. The compound is supplied as a solid of high purity, is soluble in DMSO, and is provided in small research quantities for assay controls and pathway studies. Stock solutions should be stored cold and prepared fresh for experiments.
- Selective inhibition of ALK4, ALK5, and ALK7 kinases.
- High purity suitable for biochemical and cell-based assays.
- Molecular weight 421.52 g·mol⁻¹ for accurate dosing.
- Soluble in DMSO for convenient stock preparation.
- Recommended storage of stock solution at < -20 °C and use within one month.
- Provided in small research quantities suitable for control experiments.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.