Learn More
Medchemexpress LLC 3-(6-methyl-2-pyridinyl)-N-phenyl-4-quinolin-4-ylpyrazole-1-carbothioamide | 909910-43-6 | MFCD08705403 | 99.1% | 421.52 g/mol | C25H19N5S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10432100MG
Encompass_Preferred
A small-molecule inhibitor of TGF-β type I receptors ALK5, ALK4, and ALK7 for research use. It blocks receptor-induced transcription with reported IC50 values of 12 nM (ALK5), 45 nM (ALK4), and 7.5 nM (ALK7). The product is supplied as an off-white to light yellow solid, is unstable in solution and should be prepared fresh; recommended storage is -20 °C, protected from light.
  • Potent inhibition of ALK5, ALK4, and ALK7 with low-nanomolar IC50s.
  • High purity suitable for research applications.
  • Documented solubility profiles for in vitro and in vivo use.
  • Supplied as a solid with recommended cold, light-protected storage.
  • Includes supporting documentation such as data sheet, COA, and SDS for quality assurance.
Catalog No. 50-002-43183