Learn More
Medchemexpress LLC 3-(6-methyl-2-pyridinyl)-N-phenyl-4-quinolin-4-ylpyrazole-1-carbothioamide | 909910-43-6 | MFCD08705403 | 99.1% | 421.52 g/mol | C25H19N5S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY1043225MG
Encompass_Preferred
A 83-01 is a potent small-molecule inhibitor of activin receptor-like kinases ALK4, ALK5, and ALK7 (TGF-β type I receptors), used in research to modulate TGF-β signaling in cell-based and biochemical assays. It is supplied as a solid with documented analytical data to support experimental reproducibility and quality control.
  • Potent inhibition of ALK4/5/7 for targeted pathway studies.
  • High purity suitable for biochemical and cellular assays.
  • Solid form with defined storage conditions to preserve stability.
  • Available in small milligram pack sizes for research-scale work.
  • Documented analytical data, including COA and SDS, for traceability.
Catalog No. 50-002-42602