Learn More
Medchemexpress LLC 3-(6-methylpyridin-2-yl)-N-phenyl-4-(quinolin-4-yl)-1H-pyrazole-1-carbothioamide | 909910-43-6 | MFCD08705403 | >98.0% | 421.5 Da | C25H19N5S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10432R25MG
Encompass_Preferred
A 83-01 (standard) is an analytical reference standard of a small-molecule inhibitor targeting TGF-β type I receptor (ALK5) with additional activity against ALK4 and ALK7. It is supplied as a solid research-grade reference for analytical characterization and assay development, with reported high purity and well-defined chemical properties.
Catalog No. 50-002-42513