Learn More
Medchemexpress LLC 3-(6-methylpyridin-2-yl)-N-phenyl-4-(quinolin-4-yl)-1H-pyrazole-1-carbothioamide | 909910-43-6 | MFCD08705403 | 98.7% | 421.52 g/mol | C25H19N5S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10432R100MG
Encompass_Preferred
A 83-01 (Standard) is the analytical standard of A 83-01, a selective small-molecule inhibitor of activin receptor-like kinases ALK4, ALK5, and ALK7. Supplied for research and analytical use, this reference standard is characterized for identity and purity to support assay development, QC, and compound verification.
  • Analytical standard for ALK4, ALK5, and ALK7 inhibition assays.
  • High purity suitable for analytical and research applications (reported 98.7%).
  • Characterized molecular weight 421.52 g/mol and formula C25H19N5S.
  • Supplied as a solid 100 mg quantity for laboratory use.
  • Certificate of analysis available for lot-specific purity and characterization.
  • Intended for research use only; not for human or clinical applications.
Catalog No. 50-002-43530