Learn More
Cayman Chemical 3 6DImethoxy9Hxnthen9on 1g
SDP
Supplier:  Cayman Chemical 348061 G
Encompass_Preferred
A synthetic intermediate; has been used in the synthesis of fluorescein derivatives, as well as compounds with anticancer or acetylcholinesterase inhibitory activities
Catalog No. 50-291-1178