Learn More
Medchemexpress LLC 3 7 11 15-Tetramethy 100g
SDP
Supplier:  Medchemexpress LLC HYW107616100G
Encompass_Preferred
3 7 11 15-Tetramethyl-2-hexadecen-1-ol can be used to synthesize vitamin E and vitamin E s precursor vitamin K1 3 7 11 15-Tetramethyl-2-hexadecen-1-ol regulates transcription in cells through the transcription factor PPAR-alpha and the retinoid X receptor (RXR)43[1]
Catalog No. 50-002-05579