Learn More
Medchemexpress LLC 3,7,4'-Trihydroxyflavone | 2034-65-3 | 99.4% | 270.24 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111806
Encompass_Preferred
3,7,4'-Trihydroxyflavone (5-Deoxykampferol) is a flavonoid compound isolated from the stems of Rhus javanica var. roxburghiana. It has the ability to cleave DNA in the presence of copper ions and possesses anti-inflammatory and antioxidant activities, scavenging various reactive oxygen species (ROS) and reactive nitrogen species (RNS).
  • Cleaves DNA in the presence of copper ions.
  • Exhibits anti-inflammatory activities.
  • Possesses antioxidant activities.
  • Scavenges various reactive oxygen species (ROS).
  • Scavenges reactive nitrogen species (RNS).
Catalog No. 50-004-39322