Learn More
Medchemexpress LLC 3-(7-bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione | 2229976-08-1 | MFCD32692014 | 99.0% | 323.15 g·mol⁻¹ | C13H11BrN2O3 | 5 G

Supplier: Medchemexpress LLC HYW2434055G
3-(7-Bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione is a lenalidomide analog used as a tool compound in targeted protein degradation (PROTAC) research. It is supplied as a high-purity research reagent for laboratory studies and is not intended for clinical use or sale to patients.
- High purity suitable for biochemical assays.
- Molecular formula C13H11BrN2O3 and molecular weight 323.15 g·mol⁻¹.
- Available in multiple quantities, including 5 G, for synthesis and testing.
- Storage and stability information provided for solid and solvent conditions.
- For research use only; not for human therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.