Learn More
Medchemexpress LLC 3-(7-fluoro-3-oxo-1H-isoindol-2-yl)piperidine-2,6-dione | 2359705-88-5 | MFCD31556181 | 99.9% | 262.24 g·mol⁻¹ | C13H11FN2O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW039233
Encompass_Preferred
Lenalidomide-F is a cereblon-binding E3 ligase ligand used as the recruiting moiety in PROTAC degraders targeting FLT3. It promotes ubiquitination and proteasome-dependent degradation of FLT3 in cellular studies, supporting degrader development and mechanism-of-action experiments.
  • E3 ligase ligand that binds cereblon.
  • Facilitates targeted FLT3 degradation via the ubiquitin-proteasome pathway.
  • High purity suitable for biochemical and cellular assays.
  • Stable solid with multiyear room-temperature storage.
  • Available in multiple small-scale packaging sizes for research use.
  • Molecular weight 262.24 g·mol⁻¹; formula C13H11FN2O3.
Catalog No. 50-003-03923