Learn More
AdipoGen 3 8-DAPP
SDP
Supplier:  AdipoGen CDXD0436G005
Encompass_Preferred
Chemical. CAS 52009-64-0. Formula C19H15N3. MW 285.3. Synthetic. Phenyl-phenanthridine dye with a large pi-conjugated structure and strong luminescence. Used in the synthesis of rigid polyamides and fluorescent probes. DNA intercalator. Used as a chromatographic affinity ligand for the specific separation and purification of supercoiled plasmid DNA pDNA. It has been investigated for its anti-tumor and anti-viral properties.
Catalog No. 50-256-1078