Learn More
Medchemexpress LLC 3,8-Diamino-6-phenylphenanthridine | 52009-64-0 | 285.34 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01151110G
Encompass_Preferred
3,8-Diamino-6-phenylphenanthridine is a biochemical assay reagent. It is capable of binding to DNA through a non-specific binding mechanism along the exterior of the DNA molecule, making it useful in various biochemical studies.
  • Can bind to DNA via non-specific binding
  • Classified as a biochemical assay reagent
Catalog No. 50-004-55022