Learn More
Medchemexpress LLC 3,9,15,21-tetraazatricosane-8,12,16-tricarboxylic acid (8S,16S) | 26198-65-2 | 97.2% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1222801MG
Encompass_Preferred
Aerobactin is a bacterial siderophore and virulence factor that chelates iron; it is supplied as a research-grade analytical reagent intended for in vitro studies and biochemical assays. It is provided for research use only and is commonly used to investigate iron acquisition and pathogenic mechanisms.
  • Bacterial siderophore involved in iron acquisition and pathogenicity.
  • Useful as an analytical standard in biochemical and microbiological assays.
  • High purity suitable for analytical applications (97.2%).
  • Molecular weight 564.54 g/mol and formula C22H36N4O13.
  • Supplied as a 1 mg quantity for small-scale experiments.
Catalog No. 50-002-31278