Learn More
Cayman Chemical 3-acetyl-11-ketoBoswelIc 1mg
SDP
Supplier:  Cayman Chemical 116721 MG
Encompass_Preferred
A selective 5-LO inhibitor (IC50 = 1.5 µM) that demonstrates anti-inflammatory, anti-arthritic, and anti-cancer actions
Catalog No. 50-295-1804