Learn More
Medchemexpress LLC 3-Acetylbenzophenone 250mg
SDP
Supplier:  Medchemexpress LLC 250MGHYW102471
Encompass_Preferred
3-Acetylbenzophenone (Compound 2) is the photolysis product of Ketoprofen (HY-B0227) 3-Acetylbenzophenone promotes the photoperoxidation reaction of Linoleic acid (HY-N0729) exhibits free radical scavenging and photosensitizing activity[1]
Catalog No. 50-002-04579