Learn More
Medchemexpress LLC 3-acetylindole | 703-80-0 | MFCD00005626 | 99.8% | 159.18 g/mol | C10H9NO | 50 G

Supplier: Medchemexpress LLC HYW01866750G
3-Acetylindole (CAS 703-80-0) is an acetylated indole derivative used as a synthetic intermediate in organic chemistry and as a precursor for antiviral agent synthesis. It is supplied as a solid suitable for laboratory synthesis and research-scale medicinal chemistry applications.
- High purity (99.8%).
- Molecular weight 159.18 g/mol.
- Chemical formula C10H9NO.
- Solid, white to off-white appearance.
- Useful as a synthetic intermediate and precursor in medicinal chemistry.
- Available in research-scale quantities (25-100 g and bulk options).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.