Learn More
Medchemexpress LLC (±)-3-Amino-1,2-propanediol | 616-30-8 | 91.11 | 500 G
Supplier: Medchemexpress LLC HYW002040500G
(±)-3-Amino-1,2-propanediol is a chiral raw material utilized in the synthesis of chiral vanadium (V) Schiff base complexes. It appears as a solid below 55°C and transitions to a liquid above 57°C, intended strictly for research purposes.
- Chiral raw material
- Used for synthesis of chiral vanadium (V) Schiff base complexes
- Solid below 55°C, liquid above 57°C
- For research use only