Learn More
Medchemexpress LLC 3-amino-1-(3-(1-amino-2-hydroxyethyl)-1,2,4-oxadiazol-5-yl)-3-oxopropylcarbamoyl-L-threonine | 1673534-76-3 | MFCD30738000 | 99.9% | 360.32 g/mol | C12H20N6O7 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10109350MG
Encompass_Preferred
CA-170 is an orally available small-molecule antagonist of the immune checkpoint proteins VISTA and PD-L1, used in preclinical immuno-oncology research to restore T-cell proliferation and effector functions and to evaluate checkpoint modulation strategies.
  • Orally available small-molecule inhibitor of VISTA and PD-L1.
  • Restores T-cell proliferation and effector functions in cellular assays.
  • Suitable for preclinical immuno-oncology and checkpoint modulation studies.
  • Supplied as a high-purity research reagent for analytical and in vitro use.
  • Stable when stored under recommended conditions for research purposes.
Catalog No. 50-002-11424