Learn More
Medchemexpress LLC 3-Amino-2-oxazolidinone | 80-65-9 | 98.0% | 102.09 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW01298250MG
Encompass_Preferred
3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone. It is used as an indicator of furazolidone residues in vivo and is orally active. This compound inhibits monoamine oxidase (MAO) via its metabolite in rat brain and liver.
  • Used as an indicator of furazolidone residues
  • Inhibits monoamine oxidase (MAO)
  • Increases monoamine neurotransmitter levels
  • Lowers blood pressure in renal hypertensive rats
Catalog No. 50-004-53450