Learn More
Medchemexpress LLC 3-Amino-2-oxazolidinone (AOZ) | 80-65-9 | 98.0% | 102.09 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW012982S10MG
Encompass_Preferred
3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone. It is used as an indicator for furazolidone residues in vivo and is orally active. In animal models, it has been shown to inhibit monoamine oxidase (MAO) via its metabolite in rat brain and liver, increase monoamine neurotransmitter levels (e.g., norepinephrine and dopamine) in the brain, and lower blood pressure in renal hypertensive rats.
  • Used as an indicator for furazolidone residues in vivo.
  • Orally active.
  • Inhibits monoamine oxidase (MAO).
  • Increases monoamine neurotransmitter levels in the brain.
  • Lowers blood pressure in renal hypertensive rats.
Catalog No. 50-004-52169