Learn More
Medchemexpress LLC 3-Amino-2-piperidinone | 1892-22-4 | 99.9% | 114.15 | 1 G
SDP
Supplier:  Medchemexpress LLC HY210881G
Encompass_Preferred
3-Amino-2-piperidinone is a cyclic ornithine analogue used in research to synthesize N-(2-oxopiperidin-3-yl)dodecanamide, which inhibits Fas-induced apoptosis. It is an endogenous metabolite and a delta-lactam.
  • Cyclic ornithine analogue
  • Used for synthesizing N-(2-oxopiperidin-3-yl)dodecanamide
  • Inhibits Fas-induced apoptosis
  • Suitable for laboratory research
  • Stable under recommended storage conditions
Catalog No. 50-004-01183