Learn More
Medchemexpress LLC 3-Amino-2-piperidinone | 1892-22-4 | 99.93% | 114.15 | 5 G
SDP
Supplier:  Medchemexpress LLC HY210885G
Encompass_Preferred
3-Amino-2-piperidinone is a cyclic ornithine analogue that can be used to synthesize N-(2-oxopiperidin-3-yl)dodecanamide. The synthesized N-(2-oxopiperidin-3-yl)dodecanamide is known to inhibit Fas-induced apoptosis.
  • Cyclic ornithine analogue
  • Precursor for N-(2-oxopiperidin-3-yl)dodecanamide synthesis
Catalog No. 50-004-01334