Learn More
Medchemexpress LLC 3-Amino-3-phenylpropionic acid | 614-19-7 | 165.19 | 500 G

Supplier: Medchemexpress LLC HYW002039500G
3-Amino-3-phenylpropionic acid, also known as DL-β-Phenylalanine, is a structural analogue of GABA. This compound has been observed to inhibit gastric acid secretion induced by baclofen. It appears as a white to off-white solid and is intended for research use only.
- Structural analogue of GABA
- Inhibits gastric acid secretion induced by baclofen
- White to off-white solid appearance
- Intended for research use only
- Store as powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
- Soluble in water at 5 mg/mL (requires ultrasonication)
- Insoluble or slightly soluble in DMSO (< 1 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.