Learn More
Medchemexpress LLC 3-Amino-4-hydroxybenzoic acid | 1571-72-8 | MFCD00007697 | 99.86% | 153.14 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW010224100G
Encompass_Preferred
3-Amino-4-hydroxybenzoic acid is an endogenous metabolite primarily intended for research use. It is recognized for its role as an important intermediate and a key component in pharmaceutical applications.
  • Endogenous metabolite
  • Primarily for research use
  • Key intermediate in pharmaceutical synthesis
  • Used in anti-tuberculosis drug development
  • Identified in human serum
Catalog No. 50-004-54051