Learn More
Medchemexpress LLC 3-Amino-5-bromo-2-methylpyridine | 914358-73-9 | 187.04 | 10 G

Supplier: Medchemexpress LLC HYW00357510G
3-Amino-5-bromo-2-methylpyridine (3ABMP) is a biochemical reagent. It is suitable for various biochemical assay applications.
- Used as a biochemical reagent
- Suitable for biochemical assay applications
- Molecular formula: C6H7BrN2
- Molecular weight: 187.04
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.