Learn More
Medchemexpress LLC 3-Amino-5-bromo-2-methylpyridine | 914358-73-9 | 187.04 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0035755G
Encompass_Preferred
3-Amino-5-bromo-2-methylpyridine (3ABMP) is a biochemical reagent. It has a molecular weight of 187.04 and a chemical formula of C6H7BrN2. This compound is suitable for various biochemical assay applications.
  • Suitable for biochemical assay reagents
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-004-45408