Learn More
Medchemexpress LLC 3-amino-6-chloropyridazine | 5469-69-2 | MFCD00051506 | 100.0% | 129.55 g·mol-1 | C4H4ClN3 | 100g
SDP
Supplier:  Medchemexpress LLC HYY0141100G
Encompass_Preferred
3-Amino-6-chloropyridazine is a nitrogen-containing heterocyclic organic compound belonging to the pyridazine derivatives 3-Amino-6-chloropyridazine has the basicity of amino groups and the nucleophilic substitution activity of chlorine atoms 3-Amino-6-chloropyridazine can serve as an important intermediate for synthesizing some active compounds[1][2]
Catalog No. 50-002-68879