Learn More
Medchemexpress LLC 3-amino-9-ethylcarbazole | 132-32-1 | MFCD00004964 | 99.6% | 210.28 g·mol^-1 | C14H14N2 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01438650G
Encompass_Preferred
3-Amino-9-ethylcarbazole (9-ethyl-9H-carbazol-3-amine, CAS 132-32-1) is an organic chromogenic reagent used for peroxidase-based detection in biochemical assays and immunohistochemical staining. Supplied as a high-purity solid for laboratory use, it is suitable for histology, immunoblotting, and related research applications.
  • High purity (99.61%).
  • Chromogenic substrate for peroxidase-based detection.
  • Suitable for immunohistochemistry and immunoblotting.
  • Solid form convenient for long-term storage and handling.
  • Available in multiple pack sizes for laboratory needs.
Catalog No. 50-003-03914