Learn More
Selleck Chemical LLC 3-Amino-9-ethylcarbazole E2703-100MG
SDP
Supplier:  Selleck Chemical LLC E2703100MG
Encompass_Preferred
3-Amino-9-ethylcarbazole (AEC) is a chemical compound commonly used as a chromogenic substrate in immunohistochemistry specifically for visualizing sections stained with HRP-conjugated secondary antibodies
Catalog No. 50-000-40024