Learn More
Medchemexpress LLC 3-amino-N-(2,4-dibromophenyl)-6-(pyridin-3-yl)thieno[2,3-b]pyridine-2-carboxamide | 840461-03-2 | 99.9% | 504.20 | C19H12Br2N4OS | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1381315MG
Encompass_Preferred
SOD1-Derlin-1 inhibitor-1 is a small-molecule research compound that disrupts the interaction between superoxide dismutase 1 (SOD1) and Derlin-1. It is used in preclinical and mechanistic studies of amyotrophic lateral sclerosis (ALS) to probe SOD1-mediated pathology. The compound is supplied as a light yellow solid with high reported purity for laboratory use.
  • Inhibits SOD1-Derlin-1 interaction in biochemical and cellular assays.
  • High purity suitable for research applications.
  • Light yellow solid, easy to handle and weigh.
  • Soluble in DMSO for assay preparation.
  • Available in small pack sizes suitable for lab-scale studies.
Catalog No. 50-002-22977