Learn More
Medchemexpress LLC 3-amino-N-phenyl-N-(prop-2-en-1-yl)-6-(pyridin-3-yl)thieno[2,3-b]pyridine-2-carboxamide | 2170170-27-9 | MFCD34551106 | 99.9% | 386.47 | C22H18N4OS | 5MG
SDP
Supplier:  Medchemexpress LLC HY1239825MG
Encompass_Preferred
SOD1-Derlin-1 inhibitor-2 is a small-molecule research compound that attenuates the interaction between superoxide dismutase 1 (SOD1) and Derlin-1. It is supplied for laboratory research use and is used in studies related to amyotrophic lateral sclerosis (ALS).
  • Attenuates SOD1-Derlin-1 protein-protein interaction for ALS research.
  • High purity: 99.9%.
  • Molecular formula C22H18N4OS; molecular weight 386.47.
  • Multiple supply formats, including 5 mg vials and 10 mM 1 mL in DMSO solution.
  • Certificate of analysis available online.
  • Intended for in vitro research; not for human use.
Catalog No. 50-002-35048