Learn More
Medchemexpress LLC 3-aminobenzamide | 3544-24-9 | >99.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HY120221G
Encompass_Preferred
3-aminobenzamide is a small-molecule poly(ADP-ribose) polymerase (PARP) inhibitor used in research to study DNA repair, cell death, and ischemia-reperfusion injury. It has a reported IC50 of approximately 50 nM in CHO cells.
  • Potent PARP inhibitor (IC50 ≈ 50 nM in CHO cells).
  • Used to study DNA repair and cell death pathways.
  • Mediator of oxidant-induced myocyte dysfunction during reperfusion.
  • Chemical identifiers: CAS 3544-24-9; formula C7H8N2O; MW 136.15 g/mol.
  • Commonly supplied in 1 G research quantities.
Catalog No. 50-002-31800