Learn More
Medchemexpress LLC 3-Aminobutanoic acid | 541-48-0 | 98.0% | 103.12 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00883310G
Encompass_Preferred
3-Aminobutanoic acid is a β-amino acid that can protect plants against a challenge infection with *P. infestans*. It exhibits various levels of susceptibility for the pathogen.
  • A β-amino acid
  • Can protect plants against *P. infestans* infection
  • Increases anthocyanin content in purple basil leaves
  • Reduces iron ion chelating ability
  • Enhances inhibition ability of LOX and COX-2
  • Activates phenylalanine ammonia-lyase (PAL) and polyphenol oxidase (PPO)
  • Increases mRNA expression of PAL and PPO
  • Reduces survival rate, pupation rate, and eclosion rate of tea geometrid larvae
  • Prolongs larval development period of tea geometrid
  • Induces systemic acquired resistance (SAR) in plants
  • Enhances plant resistance to pathogens and pests
Catalog No. 50-004-53424