Learn More
Medchemexpress LLC 3-Aminobutanoic acid | 541-48-0 | 98.0% | 103.12 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0088335G
Encompass_Preferred
3-Aminobutanoic acid is a β-amino acid primarily used in laboratory research. It has shown to protect plants against P. infestans infection by inducing systemic acquired resistance.
  • Exhibits various levels of susceptibility for the pathogen.
  • Increases anthocyanin content in purple basil leaves.
  • Reduces iron ion chelating ability.
  • Enhances inhibition of LOX and COX-2.
  • Activates phenylalanine ammonia-lyase (PAL) and polyphenol oxidase (PPO).
  • Increases mRNA expression of PAL and PPO.
  • Reduces survival, pupation, and eclosion rates of tea geometrid larvae.
  • Prolongs larval development.
Catalog No. 50-004-51284