Learn More
Medchemexpress LLC 3-Aminoisobutyric acid | 144-90-1 | 98.0% | 103.12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW012974R50MG
Encompass_Preferred
3-Aminoisobutyric acid (β-Aminoisobutyric acid) has anti-inflammatory and antioxidant effects. It is a catabolic metabolite of thymine and valine in skeletal muscle. This product is suitable for laboratory research applications.
  • Increases the expression of brown adipocyte-specific genes
  • Increases fatty acid β-oxidation in hepatocytes
  • Attenuates insulin resistance
  • Reduces inflammation
  • Suitable for in vitro and in vivo studies
Catalog No. 50-003-66752