Learn More
AdipoGen 3-Aminoisobutyric acid, 214139-20-5, MFCD00008145, 250 mg
SDP
Supplier:  AdipoGen AGCR13596M250
Encompass_Preferred
MF: C4H9NO2, MW: 103.120, ≥98%, Appearance: White solid. Catabolite of thymine, Precursor for 3-Hydroxyisobutyric acid synthesis, Small molecule myokine. Browning inducer of white adipose tissue (WAT) into brown adipose tissue (BAT), Induces a brown adipose-like phenotype in human pluripotent stem cells and improves glucose homeostasis.
Catalog No. 50-101-6309