Learn More
Medchemexpress LLC 3-(aminomethyl)pyridine | 3731-52-0 | MFCD00006412 | 99.4% | 108.14 | C6H8N2 | 50g
SDP
Supplier:  Medchemexpress LLC HYY004750G
Encompass_Preferred
3-Picolylamine is a primary amine compound containing a pyridine ring structure 3-Picolylamine inhibits histaminase (diamine oxidase) activity in pig kidneys 3-Picolylamine significantly enhances the contractile response of isolated guinea pig ileum to Histamine (HY-B1204) 3-Picolylamine can be used to study histamine-related physiological mechanisms or allergic diseases (such as histamine-mediated inflammatory responses)[1][2]
Catalog No. 50-002-68816