Learn More
Medchemexpress LLC 3-aminopropionitrile fumarate (2:1) | 2079-89-2 | MFCD00013142 | 99.9% | 256.26 | C4H4O4·2C3H6N2 | 10 G
SDP
Supplier:  Medchemexpress LLC HY10782910G
Encompass_Preferred
3-Aminopropionitrile fumarate (2:1) is the fumarate salt of β-aminopropionitrile supplied for laboratory research. It acts as a lathyrogen that inhibits collagen crosslinking and is used to study connective tissue remodeling, wound healing, and lysyl oxidase-related processes. The material is supplied as a solid, white to off-white powder with high purity and defined storage guidance.
  • Inhibits crosslinking of collagen.
  • Useful for studies of connective tissue remodeling and wound healing.
  • Provided as a stable fumarate salt for improved handling.
  • High purity suitable for research applications.
  • Available in multiple package sizes for experimental flexibility.
Catalog No. 50-002-42419