Learn More
Cayman Chemical 3-AmInosalIcylIc AcId 100mg
SDP
Supplier:  Cayman Chemical 36543100 MG
Encompass_Preferred
A salicylic acid derivative; a potential impurity in commercial preparations of 5-aminosalicylic acid; protects against manganese-induced apoptosis, reductions in mitochondrial complex I activity, and increases in ROS production in SK-N-MC cells at 1 µM
Catalog No. 50-291-3312