Learn More
Accela Chembio Inc 3-aminothiophene Oxalate | 5g | 478149-05-2 | 861965-63-1 | MFCD11111630 | 97+% | Shelf Life: 1980 Days | Moisture Sensitive/n2/store In Freezer(-20) Upon Receipt
Supplier:  Accela Chembio Inc SY0030225G
Encompass_Preferred
3-aminothiophene Oxalate | 5g | 478149-05-2 | 861965-63-1 | MFCD11111630 | 97+% | Shelf Life: 1980 Days | Moisture Sensitive/n2/store In Freezer(-20) Upon Receipt
Catalog No. 50-353-594