Learn More
Cayman Chemical 3 Arachidonic Acid methyl es
SDP
Supplier:  Cayman Chemical 10006454500 UG
Encompass_Preferred
A rare polyunsaturated fatty acid; a neutral, lipid-soluble form of the rare fatty acid ω-3 arachidonic acid
Catalog No. 50-289-0041