Learn More
Medchemexpress LLC 3-azabicyclo[3.1.0]hexane-2-carboxamide, N-[3-amino-1-(cyclobutylmethyl)-2,3-dioxopropyl] | 394730-60-0 | MFCD22208555 | 98.5% | 519.7 g/mol | C27H45N5O5 | 200 MG

Supplier: Medchemexpress LLC HY10237200MG
Boceprevir (CAS 394730-60-0) is a small-molecule inhibitor of the HCV NS3 protease used for biochemical and cell-based research applications. It is supplied as a research-grade solid with a high reported purity suitable for experimental assays.
- High purity suitable for research use.
- Molecular weight 519.7 g/mol.
- Chemical formula C27H45N5O5.
- Intended for biochemical and cell-based assays targeting viral protease activity.
- Available as a 200 MG pack size.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.