Learn More
Medchemexpress LLC 3-Azetidinamine, 1-(8-bromopyrido[2,3-e]tetrazolo[1,5-a]pyrazin-4-yl)-N-methyl- | 1429375-54-1 | 99.8% | 335.16 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111501
Encompass_Preferred
H4R antagonist 1 is a potent and highly selective histamine H4 receptor (H4R) antagonist with an IC50 of 27 nM. It does not show any noticeable binding affinity to other subtypes of histamine receptors (H1R, H2R, and H3R).
  • Potent and highly selective H4 receptor antagonist
  • Does not bind to H1R, H2R, and H3R
  • Shows significant antipruritic and anti-inflammatory efficacy
  • Available as a light yellow to yellow solid
  • Provides stable storage for both powder and solution
  • Soluble in various in vitro and in vivo dissolution protocols
Catalog No. 50-003-46999