Learn More
Medchemexpress LLC 3-Azetidinemethanol hydrochloride | 928038-44-2 | 123.58 | 1 G
SDP
Supplier:  Medchemexpress LLC HY213361G
Encompass_Preferred
3-Azetidinemethanol hydrochloride is an intermediate used in the synthesis of SHP2 inhibitors. It is suitable for laboratory research applications.
  • Intermediate for chemical synthesis
  • Used in SHP2 inhibitor synthesis
  • Suitable for research use
Catalog No. 50-004-02828