Learn More
Cayman Chemical 3-AzIdo-2 3-dIdeoxyurIdIn 10mg
SDP
Supplier:  Cayman Chemical 3654410 MG
Encompass_Preferred
An antiviral nucleoside analog; inhibits cytopathogenicity induced by HIV or MSV in MT-4 cells (EC50s = 0.36 and 15 µM, respectively)
Catalog No. 50-291-3841