Learn More
Medchemexpress LLC 3'-azido-2',3'-dideoxyuridine | 84472-85-5 | 99.1% | 253.21 | C9H11N5O4 | 100 MG

Supplier: Medchemexpress LLC HY106850100MG
3′-Azido-2′,3′-dideoxyuridine is a nucleoside analog and potent inhibitor of HIV replication used in biochemical research. The compound contains an azide functional group that enables copper-catalyzed and strain-promoted azide-alkyne cycloaddition reactions for labeling and conjugation. It is supplied as a white to off-white solid with high reported purity for laboratory applications.
- High purity (99.11%) suitable for research use.
- Azide functional group compatible with CuAAC and SPAAC click chemistry.
- Stable as a powder at -20°C for extended storage.
- In solvent: stable at -80°C for 6 months and -20°C for 1 month.
- White to off-white solid, easy to handle in laboratory workflows.
- Molecular formula C9H11N5O4, molecular weight 253.21.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.