Learn More
Medchemexpress LLC 3'-azido-2',3'-dideoxyuridine | 84472-85-5 | 99.1% | 253.21 | C9H11N5O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1068505MG
Encompass_Preferred
3'-Azido-2',3'-dideoxyuridine is a nucleoside analog research reagent that inhibits human immunodeficiency virus (HIV) replication in peripheral blood mononuclear cells (PBMC). It contains an azide functional group that enables copper-catalyzed or strain-promoted azide-alkyne cycloaddition for bioconjugation and labeling in chemical biology and antiviral research.
  • Inhibits HIV replication in PBMC assays.
  • Contains an azide group for copper-catalyzed and strain-promoted click chemistry (CuAAC, SPAAC).
  • High chemical purity suitable for analytical and biological studies.
  • Supplied in small milligram quantities for research use.
  • Applicable in antiviral mechanism studies and bioconjugation workflows.
Catalog No. 50-002-40752