Learn More
Medchemexpress LLC 3'-Azido-3'-deoxy-5-methylcytidine | 1282040-14-5 | 99.9% | 282.26 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11164010MM/1ML
Encompass_Preferred
3'-Azido-3'-deoxy-5-methylcytidine (CS-92) is a potent xenotropic murine leukemia-related retrovirus (XMRV) inhibitor with a CC50 of 43.5 μM in MCF-7 cells. It also inhibits HIV-1 reverse transcriptase with an EC50 of 0.06 μM in peripheral blood mononuclear (PBM) cells. This compound is a click chemistry reagent, containing an Azide group, and can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Inhibits HIV-1 at an EC50 of 0.06 μM in PBM cells
  • Inhibits XMRV at a CC50 of 43.5 μM in MCF-7 cells
  • Functions as a click chemistry reagent
  • Contains an Azide group for CuAAc reactions with Alkyne groups
  • Participates in SPAAC reactions with DBCO or BCN groups
Catalog No. 50-004-39926